8-Nitroquinoline ≥99.0% (by GC, titration analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
N0345-25G
N0345-5G
N0345-100G
TCIAN0345-25GEA
90
EUR
TCIAN0345-25G
TCIAN0345-5G
TCIAN0345-100G
8-Nitroquinoline ≥99.0% (by GC, titration analysis)
8-Nitroquinoline
Formula:
C₉H₆N₂O₂ Peso molecolare: 174,16 g/mol Punto di ebollizione: 323 °C (1013 hPa) Punto di fusione: 89…91 °C |
MDL Number:
MFCD00006806 CAS Number: 607-35-2 EINECS: 210-135-9 |
Risultati del test di specificazione
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 99.0 % |
Purity (Nonaqueous Titration) | min. 99.0 % |
Melting point | 87.0 to 91.0 deg-C |
Solubility in hot Methanol | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...