6-Nitroquinoline ≥98.0% (by GC, titration analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
N0252-25G
N0252-5G
TCIAN0252-25GEA
121
EUR
TCIAN0252-25G
TCIAN0252-5G
6-Nitroquinoline ≥98.0% (by GC, titration analysis)
6-Nitroquinoline
Formula:
C₉H₆N₂O₂ Peso molecolare: 174,16 g/mol Punto di ebollizione: 335 °C (1013 hPa) Punto di fusione: 75 °C |
MDL Number:
MFCD00006799 CAS Number: 613-50-3 EINECS: 210-346-6 |
Risultati del test di specificazione
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 150.0 to 154.0 deg-C |
Solubility in hot Methanol | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...