Quinaldic acid ≥98.0% (by HPLC, titration analysis)
Fornitore: TCI
|
Attenzione
|
Formula:
C₁₀H₇NO₂ Punto di fusione: 154…157 °C Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00006752 CAS Number: 93-10-7 EINECS: 202-218-3 Merck Index: 14,08046 |
|
Risultati del test di specificazione
Appearance | White to Orange to Green powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 156.0 to 160.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...