Stampa…

2-Bromoquinoline ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

B3524-1G B3524-5G
TCIAB3524-1GEA 64 EUR
TCIAB3524-1G TCIAB3524-5G
2-Bromoquinoline ≥98.0% (by GC, titration analysis)
2-Bromoquinoline
Formula: C₉H₆BrN
Peso molecolare: 208,06 g/mol
Punto di ebollizione: 115°C
MDL Number: MFCD00234480
CAS Number: 2005-43-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=53c0158ac0640eee6872ffd84d2dae4301e4911e&vid=76e3485a6e6c81ee1f870653114b36f794c9ec9c&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 47.0 to 51.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR