Stampa…

2,4,6-Trimethylbenzoic acid ≥98.0% (by GC, titration analysis)

Fornitore: TCI

T1421-10G T1421-25G
TCIAT1421-10GEA 37 EUR
TCIAT1421-10G TCIAT1421-25G
2,4,6-Trimethylbenzoic acid ≥98.0% (by GC, titration analysis)
2,4,6-Trimethylbenzoic acid
Formula: C₁₀H₁₂O₂
Peso molecolare: 164.2 g/mol
Punto di ebollizione: 298
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00002481
CAS Number: 480-63-7
EINECS: 207-553-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ae26bc2edb09d17c7a92d0f20666ae24e98928a7&vid=e092cb655c32b99099c83a997116a788987f06cb&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 152.0 to 156.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR