Stampa…

4-Bromothiophene-2-carbaldehyde ≥98.0% (by GC)

Fornitore: TCI EUROPE

Synonyms: 4-Bromo-2-thiophenecarboxaldehyde, 4-Bromothiophene-2-carboxaldehyde

B2827-25G B2827-5G
TCIAB2827-25GEA 128 EUR
TCIAB2827-25G TCIAB2827-5G
4-Bromothiophene-2-carbaldehyde ≥98.0% (by GC)
4-Bromothiophene-2-carbaldehyde
Formula: C₅H₃BrOS
Peso molecolare: 191,05 g/mol
Punto di ebollizione: 115°C
MDL Number: MFCD00005431
CAS Number: 18791-75-8
EINECS: 242-577-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=58a2ec96d2d08f91ded06a372711b03e5c9c0df4&vid=58a2ec96d2d08f91ded06a372711b03e5c9c0df4&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Yellow to Orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 44.0 to 48.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR