Stampa…

Benzylammonium chloride ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI
Attenzione

Synonyms: Benzylamine hydrochloride

B0407-25G B0407-500G B0407-100G
TCIAB0407-25GEA 22 EUR
TCIAB0407-25G TCIAB0407-500G TCIAB0407-100G
Benzylammonium chloride ≥98.0% (by HPLC, titration analysis)
Benzylammonium chloride
Formula: C₇H₁₀ClN
MDL Number: MFCD00012852
CAS Number: 3287-99-8
EINECS: 221-943-6
Merck Index: 14,01125
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3217ee53a136c503f80a5849be165c435c7840c7&vid=3217ee53a136c503f80a5849be165c435c7840c7&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Solubility in Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR