Boc-Pro-OH ≥99.0% (by titrimetric analysis)
Fornitore: TCI EUROPE
Synonyms:
N-(tert-Butoxycarbonyl)-L-proline, N-Boc-L-proline, N-α-t-Boc-L-Proline, BOC-L-Proline, (S)-1-(tert-Butoxycarbonyl)pyrrolidine-2-carboxylic acid
B1188-25G
B1188-5G
TCIAB1188-25GEA
43
EUR
TCIAB1188-25G
TCIAB1188-5G
Boc-Pro-OH ≥99.0% (by titrimetric analysis)
Boc-Pro-OH
Formula:
C₁₀H₁₇NO₄ Peso molecolare: 215,25 g/mol |
MDL Number:
MFCD00037324 CAS Number: 15761-39-4 EINECS: 239-848-3 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Optical purity (LC) | min. 98.0 ee% |
Purity (Neutralization titration) | min. 99.0 % |
Melting point | 132.0 to 137.0 deg-C |
Specific rotation [a]20/D | -58.0 to -62.0 deg (C=2, AcOH) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...