Stampa…

Boc-D-Pro-OH ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI EUROPE

Synonyms: (2R)-1-[(tert-butoxy)carbonyl]pyrrolidine-2-carboxylic acid, N-Boc-D-proline

B2977-25G B2977-5G
TCIAB2977-25GEA 37 EUR
TCIAB2977-25G TCIAB2977-5G
Boc-D-Pro-OH ≥98.0% (by HPLC, titration analysis)
Boc-D-Pro-OH
Formula: C₁₀H₁₇NO₄
Peso molecolare: 215,25 g/mol
MDL Number: MFCD00063226
CAS Number: 37784-17-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=db32a77a2c50f25ddd1f39b656dbf99f7fa0fd3d&vid=db32a77a2c50f25ddd1f39b656dbf99f7fa0fd3d&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 132.0 to 137.0 deg-C
Specific rotation [a]20/D +58.0 to +62.0 deg (C=2, AcOH)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR