Stampa…

Farnesol (mixture of isomers)

Fornitore: TCI EUROPE
Attenzione

Synonyms: Farnesol, 3,7,11-Trimethyldodeca-2,6,10-trien-1-ol, TRANS,TRANS-3,7,11-TRIMETHYL-2,6,10-DODECATRIEN-1-OL, 2,6,10-Dodecatrien-1-ol, 3,7,11-trimethyl-, (2Z,6Z)-

T0608-25G T0608-5G T0608-100G
TCIAT0608-25GEA 31 EUR
TCIAT0608-25G TCIAT0608-5G TCIAT0608-100G
Farnesol (mixture of isomers)
Farnesol (mixture of isomers)
Formula: C₁₅H₂₆O
Peso molecolare: 222,37 g/mol
Punto di ebollizione: 143°C
Flash Pt: 96°C
MDL Number: MFCD00002918
CAS Number: 4602-84-0
EINECS: 225-004-1
Merck Index: 14,03937
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=60a4bcb2abeedc288b290f286eba6e40c6e4acf7&vid=60a4bcb2abeedc288b290f286eba6e40c6e4acf7&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Brown clear liquid
Purity (GC) min. 95.0 % (total of isomer)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR