Methyldiphenylamine ≥98.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
N-Methyl-N-phenylaniline
M0211-25ML
M0211-5ML
TCIAM0211-25MLEA
86
EUR
TCIAM0211-25ML
TCIAM0211-5ML
Methyldiphenylamine ≥98.0% (by GC)
Methyldiphenylamine
Formula:
C₁₃H₁₃N Peso molecolare: 183,25 g/mol Punto di ebollizione: 295°C |
MDL Number:
MFCD00041900 CAS Number: 552-82-9 EINECS: 209-023-2 Merck Index: 14,06054 |
Risultati del test di specificazione
Appearance | Light yellow to Yellow clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...