(S,S)-(-)-Hydrobenzoin ≥99.0% (by GC)
Fornitore: TCI EUROPE
Synonyms:
(1S,2S)-1,2-Diphenylethane-1,2-diol
H0824-25G
H0824-5G
TCIAH0824-25GEA
556
EUR
TCIAH0824-25G
TCIAH0824-5G
(S,S)-(-)-Hydrobenzoin ≥99.0% (by GC)
(S,S)-(-)-Hydrobenzoin
Formula:
C₁₄H₁₄O₂ Peso molecolare: 214,26 g/mol |
MDL Number:
MFCD00064255 CAS Number: 2325-10-2 Merck Index: 14,04777 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 99.0 % |
Optical purity (LC) | min. 98.0 ee% |
Melting point | 146.0 to 150.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...