Stampa…

1-(4-Bromophenyl)ethylamine ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Pericolo

Synonyms: 4-Bromo-α-methylbenzylamine, (+/-)-1-(4-Bromophenyl)ethylamine, (±)-4-Bromo-α-methylbenzylamine

B2266-25G B2266-5G
TCIAB2266-25GEA 307 EUR
TCIAB2266-25G TCIAB2266-5G
1-(4-Bromophenyl)ethylamine ≥98.0% (by GC, titration analysis)
1-(4-Bromophenyl)ethylamine
Formula: C₈H₁₀BrN
Peso molecolare: 200,08 g/mol
MDL Number: MFCD00025548
CAS Number: 24358-62-1
EINECS: 246-200-3
UN: 2735
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=22595e5c1b0b535339496af8c13e9f7157ecd560&vid=22595e5c1b0b535339496af8c13e9f7157ecd560&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR