tert-Butylchlorodiphenylsilane ≥97.0% (by GC)
Fornitore: TCI EUROPE
|
Pericolo
|
Synonyms:
tert-Butyldiphenylchlorosilane, tert-Butyl-diphenylchlorosilane
B1223-100ML
B1223-25ML
B1223-5ML
TCIAB1223-100MLEA
243
EUR
TCIAB1223-100ML
TCIAB1223-25ML
TCIAB1223-5ML
tert-Butylchlorodiphenylsilane ≥97.0% (by GC)
tert-Butylchlorodiphenylsilane
Formula:
C₁₆H₁₉ClSi Peso molecolare: 274,87 g/mol Punto di ebollizione: 125°C Flash Pt: 112°C |
MDL Number:
MFCD00000497 CAS Number: 58479-61-1 EINECS: 261-282-0 UN: 2987 ADR: 8,II |
Risultati del test di specificazione
Appearance | Colorless to Light orange to Yellow clear liquid |
Purity (GC) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...