Stampa…

Ethanolamine hydrochloride ≥98.0% (by titrimetric analysis)

Fornitore: TCI
Attenzione

Synonyms: 2-Aminoethanol hydrochloride, 2-Hydroxyethylammonium Chloride, Monoethanolamine Hydrochloride, 2-Aminoethan-1-ol hydrochloride

A0298-25G A0298-500G A0298-100G
TCIAA0298-25GEA 17 EUR
TCIAA0298-25G TCIAA0298-500G TCIAA0298-100G
Ethanolamine hydrochloride ≥98.0% (by titrimetric analysis)
Ethanolamine hydrochloride
Formula: C₂H₈ClNO
Peso molecolare: 97.54 g/mol
MDL Number: MFCD00012892
CAS Number: 2002-24-6
EINECS: 217-900-6
Merck Index: 14,03727
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7813253f0c90bd4375e654d764e68dec7872d2f2&vid=7813253f0c90bd4375e654d764e68dec7872d2f2&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White powder to crystal
Purity (Argentmetric Titration) min. 98.0 %
Melting point 82.0 to 86.0 deg-C
Solubility in Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR