Stampa…

8-Amino-1-naphthalenesulphonic acid ≥98.0% (by HPLC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 8-Aminonaphthalene-1-sulphonic acid

A0350-25G A0350-100G
TCIAA0350-25GEA 23 EUR
TCIAA0350-25G TCIAA0350-100G
8-Amino-1-naphthalenesulphonic acid ≥98.0% (by HPLC)
8-Amino-1-naphthalenesulphonic acid
Formula: C₁₀H₉NO₃S
Peso molecolare: 223,25 g/mol
MDL Number: MFCD00035730
CAS Number: 82-75-7
EINECS: 201-437-1
Merck Index: 14,06405
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=48c398d65a74796e6be83169ea1713256f33c83e&vid=48c398d65a74796e6be83169ea1713256f33c83e&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Light yellow to Brown powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 95.0 %
Solubility in NH3aq. almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR