Stampa…
2,2-Diphenylethylamine ≥98.0% (by GC, titration analysis)
Fornitore: TCI EUROPE
Attenzione
Synonyms: 1-Amino-2,2-diphenylethane, 2,2-Diphenylethanamine
D2018-25G D2018-5G
TCIAD2018-25GEA 220 EUR
TCIAD2018-25G TCIAD2018-5G
2,2-Diphenylethylamine ≥98.0% (by GC, titration analysis)
2,2-Diphenylethylamine
Formula: C₁₄H₁₅N
Peso molecolare: 197,28 g/mol
MDL Number: MFCD00008143
CAS Number: 3963-62-0
EINECS: 223-565-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0ade8be3aa926e8aeca1a365c3dc769cf364eee3&vid=0ade8be3aa926e8aeca1a365c3dc769cf364eee3&prid=web&clid=SAPDAM
Visualizza i codici
Appearance White to Light yellow to Light orange powder to lump
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 45.0 to 49.0 deg-C