Stampa…
4-Bromo-2-thenoic acid ≥98.0% (by GC, titration analysis)
Fornitore: TCI EUROPE
Attenzione
Synonyms: 4-Bromo-2-thiophenecarboxylic acid
B3386-25G B3386-5G
TCIAB3386-25GEA 221 EUR
TCIAB3386-25G TCIAB3386-5G
4-Bromo-2-thenoic acid ≥98.0% (by GC, titration analysis)
4-Bromo-2-thenoic acid
Formula: C₅H₃BrO₂S
Peso molecolare: 207,05 g/mol
MDL Number: MFCD03422294
CAS Number: 16694-18-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c05f44564d653c09786d2242041ca1a26975c026&vid=c05f44564d653c09786d2242041ca1a26975c026&prid=web&clid=SAPDAM
Visualizza i codici
Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 120.0 to 124.0 deg-C