Acido-3-amino-benzensolfonico ≥99.0% (by titrimetric analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
1-Aminobenzene-3-sulphonic acid, Metanilic acid, 3-Aminobenzenesulphonic acid
A0265-25G
A0265-500G
TCIAA0265-25GEA
16
EUR
TCIAA0265-25G
TCIAA0265-500G
Acido-3-amino-benzensolfonico ≥99.0% (by titrimetric analysis)
Acido-3-amino-benzensolfonico
Formula:
C₆H₇NO₃S Peso molecolare: 173,19 g/mol |
MDL Number:
MFCD00065345 CAS Number: 121-47-1 EINECS: 204-473-6 Merck Index: 14,05927 |
Risultati del test di specificazione
Appearance | White to Light yellow to Dark green powder to crystal |
Purity (HPLC) | min. 97.0 area% |
Purity (Neutralization titration) | min. 99.0 % |
Solubility in hot Water | very faint turbidity |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...