Stampa…

4-Chlorophenethylamine ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 2-(4-Chlorophenyl)ethylamine

C1129-25G C1129-5G
TCIAC1129-25GEA 79 EUR
TCIAC1129-25G TCIAC1129-5G
4-Chlorophenethylamine ≥98.0% (by GC, titration analysis)
4-Chlorophenethylamine
Formula: C₈H₁₀ClN
Peso molecolare: 155,63 g/mol
Punto di ebollizione: 140°C
Flash Pt: 106°C
MDL Number: MFCD00008191
CAS Number: 156-41-2
EINECS: 205-853-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9cb7b5795e1d71fc99de9b6f032ddc03c080a9a3&vid=9cb7b5795e1d71fc99de9b6f032ddc03c080a9a3&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR