3,4-Dichlorophenethylamine ≥98.0% (by GC, titration analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
D2927-1G
D2927-5G
TCIAD2927-1GEA
38
EUR
TCIAD2927-1G
TCIAD2927-5G
3,4-Dichlorophenethylamine ≥98.0% (by GC, titration analysis)
3,4-Dichlorophenethylamine
Formula:
C₈H₉Cl₂N Peso molecolare: 190,07 g/mol |
MDL Number:
MFCD00060617 CAS Number: 21581-45-3 |
Risultati del test di specificazione
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...