Stampa…

3-Methoxyphenethylamine ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Pericolo

Synonyms: 2-(3-Methoxyphenyl)ethylamine

M1891-25G M1891-5G
TCIAM1891-25GEA 103 EUR
TCIAM1891-25G TCIAM1891-5G
3-Methoxyphenethylamine ≥98.0% (by GC, titration analysis)
3-Methoxyphenethylamine
Formula: C₉H₁₃NO
Peso molecolare: 151,21 g/mol
Punto di ebollizione: 119°C
MDL Number: MFCD00008187
CAS Number: 2039-67-0
EINECS: 218-017-9
UN: 2735
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2fb231555fde14b9e58c904f87b79cd28cbaeb0b&vid=2fb231555fde14b9e58c904f87b79cd28cbaeb0b&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR