Stampa…

2-Methoxyphenethylamine ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Pericolo

M0844-25ML M0844-5ML
TCIAM0844-25MLEA 115 EUR
TCIAM0844-25ML TCIAM0844-5ML
2-Methoxyphenethylamine ≥98.0% (by GC, titration analysis)
2-Methoxyphenethylamine
Formula: C₉H₁₃NO
Peso molecolare: 151,21 g/mol
Punto di ebollizione: 237°C
MDL Number: MFCD00008186
CAS Number: 2045-79-6
EINECS: 218-066-6
UN: 3267
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=31899be9bebf37ee56e7066a654ae5c4e5ab8699&vid=31899be9bebf37ee56e7066a654ae5c4e5ab8699&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White or Colorless to Light yellow powder to lump to clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR