Stampa…

Benzoin ≥98.0% (by GC)

Fornitore: TCI EUROPE

Synonyms: 2-Hydroxy-2-phenylacetophenone

B0079-25G B0079-500G
TCIAB0079-25GEA 16 EUR
TCIAB0079-25G TCIAB0079-500G
Benzoin ≥98.0% (by GC)
Benzoin
Formula: C₁₄H₁₂O₂
Peso molecolare: 212,25 g/mol
MDL Number: MFCD00004496
CAS Number: 119-53-9
EINECS: 204-331-3
Merck Index: 14,01093
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d4630446783653e9e35a05fd1d6d43f79994bdda&vid=d4630446783653e9e35a05fd1d6d43f79994bdda&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 133.0 to 138.0 deg-C
Solubility in hot Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR