Stampa…

Benzoin methyl ether ≥98.0% (by GC)

Fornitore: TCI

Synonyms: 1,2-Diphenyl-2-methoxyethan-1-one, 2-Methoxy-1,2-diphenylethanone

B0869-25G B0869-500G
TCIAB0869-25GEA 40 EUR
TCIAB0869-25G TCIAB0869-500G
Benzoin methyl ether ≥98.0% (by GC)
Benzoin methyl ether
Formula: C₁₅H₁₄O₂
Punto di ebollizione: 189°C
MDL Number: MFCD00008492
CAS Number: 3524-62-7
EINECS: 222-538-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d4f8bfb7b35faa3b64d8d71b05258f7e603b05db&vid=d4f8bfb7b35faa3b64d8d71b05258f7e603b05db&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 46.0 to 52.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR