Stampa…

Benzoin isopropyl ether ≥99.0% (by GC)

Fornitore: TCI EUROPE

Synonyms: 2-Isopropoxy-1,2-diphenylethanone

B0942-25G B0942-500G
TCIAB0942-25GEA 24 EUR
TCIAB0942-25G TCIAB0942-500G
Benzoin isopropyl ether ≥99.0% (by GC)
Benzoin isopropyl ether
Formula: C₁₇H₁₈O₂
Peso molecolare: 254,33 g/mol
Punto di ebollizione: 180°C
MDL Number: MFCD00015025
CAS Number: 6652-28-4
EINECS: 229-677-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=638ff9d1c268adee2c3c047e2e85ae20ca37434b&vid=638ff9d1c268adee2c3c047e2e85ae20ca37434b&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 99.0 %
Melting point 78.0 to 82.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR