Desyl chloride ≥98.0% (by GC)
Fornitore: TCI EUROPE
|
Pericolo
|
Synonyms:
2-Chloro-2-phenylacetophenone
D1745-25G
D1745-5G
TCIAD1745-25GEA
81
EUR
TCIAD1745-25G
TCIAD1745-5G
Desyl chloride ≥98.0% (by GC)
Desyl chloride
Formula:
C₁₄H₁₁ClO Peso molecolare: 230,69 g/mol |
MDL Number:
MFCD00000858 CAS Number: 447-31-4 EINECS: 207-181-7 UN: 3261 ADR: 8,II |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 65.0 to 70.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...