Stampa…

(1-Chloroethyl)benzene ≥97.0% (by GC)

Fornitore: TCI

Synonyms: 1-Phenylethyl chloride

C0166-25ML C0166-500ML
TCIAC0166-25MLEA 43 EUR
TCIAC0166-25ML TCIAC0166-500ML
(1-Chloroethyl)benzene ≥97.0% (by GC)
(1-Chloroethyl)benzene
Formula: C₆H₅CHClCH₃
Peso molecolare: 140.61 g/mol
MDL Number: MFCD00018870
CAS Number: 672-65-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1824dddceabd3309f2a844e5d9d7b104a7687d7c&vid=889b140010566792a17e4bf18bdddb7f87fccd93&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR