4-Hydroxy-3-methylbenzoic acid ≥98.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
4-Hydroxy-m-toluic acid
H1319-25G
H1319-5G
TCIAH1319-25GEA
190
EUR
TCIAH1319-25G
TCIAH1319-5G
4-Hydroxy-3-methylbenzoic acid ≥98.0% (by GC)
4-Hydroxy-3-methylbenzoic acid
Formula:
C₈H₈O₃ Peso molecolare: 152,15 g/mol Punto di fusione: 175 °C |
MDL Number:
MFCD00270105 CAS Number: 499-76-3 EINECS: 207-890-1 |
Risultati del test di specificazione
Appearance | Light yellow to Yellow to Orange powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 95.0 % |
Melting point | 173.0 to 177.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...