Stampa…

3-Hydroxy-p-toluic acid ≥98.0% (by GC, titration analysis)

Fornitore: TCI
Attenzione

Synonyms: 3-Hydroxy-4-methylbenzoic acid

H0929-25G H0929-5G
TCIAH0929-25GEA 187 EUR
TCIAH0929-25G TCIAH0929-5G
3-Hydroxy-p-toluic acid ≥98.0% (by GC, titration analysis)
3-Hydroxy-p-toluic acid
Formula: C₈H₈O₃
Punto di ebollizione: 313 °C (1013 hPa)
Punto di fusione: 207…209 °C
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00002511
CAS Number: 586-30-1
EINECS: 209-571-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=984d440b23900484cff0ff66e65901c8489943cf&vid=4abd8a2c26e5d6796ae2c1ab58b42d235beedda3&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 209.0 to 212.0 deg-C
Solubility in hot Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR