Stampa…

2-Hydroxy-p-toluic acid ≥99.0% (by HPLC)

Fornitore: TCI
Attenzione

C0409-25G C0409-500G
TCIAC0409-25GEA 27 EUR
TCIAC0409-25G TCIAC0409-500G
2-Hydroxy-p-toluic acid ≥99.0% (by HPLC)
2-Hydroxy-p-toluic acid
Formula: C₈H₈O₃
Punto di fusione: 177 °C
MDL Number: MFCD00002454
CAS Number: 50-85-1
EINECS: 200-068-3
Merck Index: 14,02580
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e3b7ce25215e390a9099abd9a4af9dcbdc628b9d&vid=d63f0994cf3a59c3072ae71d3ae621d85d7396c0&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow to Light orange powder to crystal
Purity (HPLC) min. 99.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 177.0 to 180.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR