4-Amino-m-toluic acid ≥98.0% (by HPLC, titration analysis)
Fornitore: TCI
|
Attenzione
|
Formula:
C₈H₉NO₂ Punto di ebollizione: 340 °C (1013 hPa) Punto di fusione: 170 °C |
MDL Number:
MFCD00007736 CAS Number: 2486-70-6 EINECS: 219-629-9 |
|
Risultati del test di specificazione
Appearance | White to Light yellow to Light red powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 170.0 to 174.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...