5-Methylanthranil acid ≥98.0% (by HPLC, titration analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
2-Amino-5-methylbenzoic acid, 6-Amino-m-toluic acid
A1421-25G
A1421-5G
TCIAA1421-25GEA
120
EUR
TCIAA1421-25G
TCIAA1421-5G
5-Methylanthranil acid ≥98.0% (by HPLC, titration analysis)
5-Methylanthranil acid
Formula:
C₈H₉NO₂ Peso molecolare: 151,16 g/mol Punto di ebollizione: 317 °C (1013 hPa) Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00007909 CAS Number: 2941-78-8 EINECS: 220-932-3 |
Risultati del test di specificazione
Appearance | White to Brown powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...