α-Bromo-3,4-difluorotoluene ≥98.0% (by GC)
Fornitore: TCI
|
Pericolo
|
Synonyms:
3,4-Difluorobenzyl bromide, 4-(Bromomethyl)-1,2-difluorobenzene
Formula:
C₇H₅BrF₂ Peso molecolare: 207.02 g/mol Punto di ebollizione: 61°C Flash Pt: 79°C |
MDL Number:
MFCD00009889 CAS Number: 85118-01-0 EINECS: 285-653-1 UN: 3265 ADR: 8 III |
|
Risultati del test di specificazione
Appearance | Colorless to Red to Green clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...