Stampa…

1,4,7,10-Tetraazaciclododecano ≥97.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Pericolo

Synonyms: Cyclen

T1874-1G T1874-5G
TCIAT1874-1GEA 99 EUR
TCIAT1874-1G TCIAT1874-5G
1,4,7,10-Tetraazaciclododecano ≥97.0% (by GC, titration analysis)
1,4,7,10-Tetraazaciclododecano
Formula: C₈H₂₀N₄
Peso molecolare: 172,27 g/mol
MDL Number: MFCD00066281
CAS Number: 294-90-6
EINECS: 425-450-9
UN: 3259
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c0243bf52a0f7189de2c0301c74527fa529ac712&vid=c0243bf52a0f7189de2c0301c74527fa529ac712&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 97.0 %
Purity (Neutralization titration) min. 97.0 %
Melting point 110.0 to 114.0 deg-C
Solubility in Water almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR