Stampa…

1,4-Diethoxybenzene ≥98.0% (by GC)

Fornitore: TCI EUROPE

D0535-25G D0535-500G
TCIAD0535-25GEA 16 EUR
TCIAD0535-25G TCIAD0535-500G
1,4-Diethoxybenzene ≥98.0% (by GC)
1,4-Diethoxybenzene
Formula: C₁₀H₁₄O₂
Peso molecolare: 166,22 g/mol
Punto di ebollizione: 112°C
MDL Number: MFCD00015146
CAS Number: 122-95-2
EINECS: 204-585-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=17e897f2681372e204da3f6882f1f8631d461add&vid=17e897f2681372e204da3f6882f1f8631d461add&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 69.0 to 73.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR