Stampa…

3-Hydroxyphenylboronic acid pinacol ester ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenol

T1953-1G T1953-5G
TCIAT1953-1GEA 55 EUR
TCIAT1953-1G TCIAT1953-5G
3-Hydroxyphenylboronic acid pinacol ester ≥98.0% (by GC)
3-Hydroxyphenylboronic acid pinacol ester
Formula: C₁₂H₁₇BO₃
Peso molecolare: 220,08 g/mol
MDL Number: MFCD02093755
CAS Number: 214360-76-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a2f9c22cfcec9a6c0f7f48ed5b94ca92093b0967&vid=a2f9c22cfcec9a6c0f7f48ed5b94ca92093b0967&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Gray to Brown powder to crystal
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR