Stampa…

4'-Hydroxybiphenyl-4-carboxylic acid ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 4'-Hydroxy-4-biphenylcarboxylic acid, 4'-Hydroxy-[1,1'-biphenyl]-4-carboxylic acid

H0695-25G H0695-5G
TCIAH0695-25GEA 240 EUR
TCIAH0695-25G TCIAH0695-5G
4'-Hydroxybiphenyl-4-carboxylic acid ≥98.0% (by GC, titration analysis)
4'-Hydroxybiphenyl-4-carboxylic acid
Formula: C₁₃H₁₀O₃
Peso molecolare: 214,22 g/mol
MDL Number: MFCD00059078
CAS Number: 58574-03-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cbec1cda0276619c53791d5335348424f9bad0d9&vid=cbec1cda0276619c53791d5335348424f9bad0d9&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Solubility in hot Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR