β-Ionone ≥95.0% (by GC)
Fornitore: TCI EUROPE
Synonyms:
4-(2,6,6-Trimethylcyclohex-1-en-1-yl)but-3-en-2-one, 4-(2,6,6-Trimethyl-1-cyclohexenyl)-3-buten-2-one
I0077-500ML
I0077-25ML
I0077-100ML
TCIAI0077-500MLEA
151
EUR
TCIAI0077-500ML
TCIAI0077-25ML
TCIAI0077-100ML
β-Ionone ≥95.0% (by GC)
β-Ionone
Formula:
C₁₃H₂₀O Peso molecolare: 192,30 g/mol Punto di ebollizione: 151°C Flash Pt: 112°C |
MDL Number:
MFCD00167092 CAS Number: 14901-07-6 EINECS: 238-969-9 Merck Index: 14,05056 |
Risultati del test di specificazione
Appearance | Light orange to Yellow to Green clear liquid |
Purity (GC) | min. 95.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...