Stampa…

(S)-(-)-1-(2-Naphthyl)ethylamine ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: (S)-1-(Naphthalen-2-yl)ethanamine

N0726-1G N0726-5G
TCIAN0726-1GEA 34 EUR
TCIAN0726-1G TCIAN0726-5G
(S)-(-)-1-(2-Naphthyl)ethylamine ≥98.0% (by GC, titration analysis)
(S)-(-)-1-(2-Naphthyl)ethylamine
Formula: C₁₂H₁₃N
Peso molecolare: 171,24 g/mol
Punto di ebollizione: 143°C
Punto di fusione: 53°C
MDL Number: MFCD00085366
CAS Number: 3082-62-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=72ee959d6485cc3172801e4dd86a17dec780b6d2&vid=72ee959d6485cc3172801e4dd86a17dec780b6d2&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Optical purity (LC) min. 98.0 ee%
Purity (Nonaqueous Titration) min. 98.0 %
Specific rotation [a]20/D -19.0 to -23.0 deg (C=1, methanol)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR