Stampa…

2-Chloro-6-methoxybenzothiazole ≥96.0% (by GC)

Fornitore: TCI

Synonyms: 2-Chloro-6-methoxy-1,3-benzothiazole, 2-Chloro-6-methoxybenzo[d]thiazole

C1464-1G C1464-5G
TCIAC1464-1GEA 29 EUR
TCIAC1464-1G TCIAC1464-5G
2-Chloro-6-methoxybenzothiazole ≥96.0% (by GC)
2-Chloro-6-methoxybenzothiazole
Formula: C₈H₆ClNOS
Punto di ebollizione: 133°C
MDL Number: MFCD00671510
CAS Number: 2605-14-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a432b9f952362c06b99d5209d02707be75a7a98f&vid=a432b9f952362c06b99d5209d02707be75a7a98f&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 96.0 %
Melting point 52.0 to 56.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR