Stampa…

Gramine ≥98.0% (by titrimetric analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 1-(1H-Indol-3-yl)-N,N-dimethylmethanamine, 3-(Dimethylaminomethyl)-Indole

D0653-25G D0653-250G
TCIAD0653-25GEA 23 EUR
TCIAD0653-25G TCIAD0653-250G
Gramine ≥98.0% (by titrimetric analysis)
Gramine
Formula: C₁₁H₁₄N₂
Peso molecolare: 174,25 g/mol
MDL Number: MFCD00005629
CAS Number: 87-52-5
EINECS: 201-749-8
Merck Index: 14,04533
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8ec9153996aef6674237ca86bcc0360d96abd356&vid=8ec9153996aef6674237ca86bcc0360d96abd356&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow to Light orange powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 131.0 to 134.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR