Stampa…

Methyl indole-6-carboxylate ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: Methyl 1H-indole-6-carboxylate

M2042-1G M2042-25G M2042-5G
TCIAM2042-1GEA 27 EUR
TCIAM2042-1G TCIAM2042-25G TCIAM2042-5G
Methyl indole-6-carboxylate ≥98.0% (by GC)
Methyl indole-6-carboxylate
Formula: C₁₀H₉NO₂
Peso molecolare: 175,19 g/mol
MDL Number: MFCD00211063
CAS Number: 50820-65-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2a2ff9672f9402f65cd1a225a08161e253b5336a&vid=2a2ff9672f9402f65cd1a225a08161e253b5336a&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR