Methyl indole-5-carboxylate ≥98.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
Methyl 1H-indole-5-carboxylate
I0545-1G
I0545-5G
TCIAI0545-1GEA
24
EUR
TCIAI0545-1G
TCIAI0545-5G
Methyl indole-5-carboxylate ≥98.0% (by GC)
Methyl indole-5-carboxylate
Formula:
C₁₀H₉NO₂ Peso molecolare: 175,19 g/mol |
MDL Number:
MFCD00153023 CAS Number: 1011-65-0 |
Risultati del test di specificazione
Appearance | White to Gray to Brown powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 125.0 to 129.0 deg-C |
Solubility in hot Methanol | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...