Stampa…

Dianidride 3,3',4,4'-benzofenontetracarbossilica ≥96.0% (by titrimetric analysis)

Fornitore: TCI
Attenzione

B0948-25G B0948-500G B0948-100G
TCIAB0948-25GEA 51 EUR
TCIAB0948-25G TCIAB0948-500G TCIAB0948-100G
Dianidride 3,3',4,4'-benzofenontetracarbossilica ≥96.0% (by titrimetric analysis)
Dianidride 3,3',4,4'-benzofenontetracarbossilica
Formula: C₁₇H₆O₇
MDL Number: MFCD00005923
CAS Number: 2421-28-5
EINECS: 219-348-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8af0a3d2f70ede3bf0248867363c5f85ff571e6c&vid=8af0a3d2f70ede3bf0248867363c5f85ff571e6c&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Orange to Green powder to crystal
Purity (Morpholine Method) min. 96.0 %
Melting point 223.0 to 229.0 deg-C
Solubility in hot Acetone very faint turbidity


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR