Stampa…

Diethyl ethoxymethylenemalonate ≥98.0% (by GC)

Fornitore: TCI
Attenzione

Synonyms: Diethyl 2-(ethoxymethylene)malonate

E0255-25ML E0255-500ML E0255-100ML
TCIAE0255-25MLEA 16 EUR
TCIAE0255-25ML TCIAE0255-500ML TCIAE0255-100ML
Diethyl ethoxymethylenemalonate ≥98.0% (by GC)
Diethyl ethoxymethylenemalonate
Formula: C₁₀H₁₆O₅
Punto di ebollizione: 166°C
Flash Pt: 155°C
MDL Number: MFCD00009148
CAS Number: 87-13-8
EINECS: 201-725-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=059d9b4e53d344ced383f142e14b1b051589b478&vid=059d9b4e53d344ced383f142e14b1b051589b478&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR