Stampa…

Diethyl fluoromalonate ≥95.0% (by GC)

Fornitore: TCI
Pericolo

Synonyms: 1,3-diethyl 2-fluoropropanedioate

F0283-1G F0283-25G F0283-5G
TCIAF0283-1GEA 23 EUR
TCIAF0283-1G TCIAF0283-25G TCIAF0283-5G
Diethyl fluoromalonate ≥95.0% (by GC)
Diethyl fluoromalonate
Formula: C₇H₁₁FO₄
Punto di ebollizione: 122°C
Flash Pt: 62°C
MDL Number: MFCD00009139
CAS Number: 685-88-1
EINECS: 211-684-7
UN: 1760
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d0ffb2f2ad9011a30239e33d6788d58b4401a19b&vid=d0ffb2f2ad9011a30239e33d6788d58b4401a19b&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR