Stampa…

2-Bromo-5-chloroaniline ≥97.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

B3182-1G B3182-25G B3182-5G
TCIAB3182-1GEA 17 EUR
TCIAB3182-1G TCIAB3182-25G TCIAB3182-5G
2-Bromo-5-chloroaniline ≥97.0% (by GC, titration analysis)
2-Bromo-5-chloroaniline
Formula: C₆H₅BrClN
Punto di ebollizione: 128°C
MDL Number: MFCD00672942
CAS Number: 823-57-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=11ab1ec5bab727bc8ac7a1a4609900da4b8a1dfb&vid=11ab1ec5bab727bc8ac7a1a4609900da4b8a1dfb&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Brown powder to lump
Purity (GC) min. 97.0 %
Purity (Nonaqueous Titration) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR