Stampa…

Diglycolic acid ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 2,2'-Oxydiacetic acid

D0545-25G D0545-500G
TCIAD0545-25GEA 31 EUR
TCIAD0545-25G TCIAD0545-500G
Diglycolic acid ≥98.0% (by GC, titration analysis)
Diglycolic acid
Formula: C₄H₆O₅
Peso molecolare: 134,09 g/mol
MDL Number: MFCD00004309
CAS Number: 110-99-6
EINECS: 203-823-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=06d7ef1cedd7782c4631df8573279fd4c3b00ff2&vid=06d7ef1cedd7782c4631df8573279fd4c3b00ff2&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 140.0 to 145.0 deg-C
Solubility in Water almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR