Stampa…

1-Methyl-3-octylimidazolium hexafluorophosphate ≥98.0% (by HPLC, total nitrogen)

Fornitore: TCI EUROPE
Attenzione

M2063-25G M2063-5G
TCIAM2063-25GEA 221 EUR
TCIAM2063-25G TCIAM2063-5G
1-Methyl-3-octylimidazolium hexafluorophosphate ≥98.0% (by HPLC, total nitrogen)
1-Methyl-3-octylimidazolium hexafluorophosphate
Formula: C₁₂H₂₃F₆N₂P
Peso molecolare: 340,29 g/mol
MDL Number: MFCD06798203
CAS Number: 304680-36-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=252827a1141f7b496b3a0241a56c77880ce10f2b&vid=7989543d9c896b665a9292f8cca1fb7002eed4c5&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Light orange to Yellow clear liquid
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR